ENA321761977 Enamine

ethyl[2-(thiophen-2-yl)ethyl]amine

  • MDL number MFCD11145624
  • Cas Number : 753413-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-32973

Safety Information

msds https://www.enaminestore.com/catalog/EN300-32973
Availability details for
Quantity
  • Availability
  • Add to Cart