ENA321761982 Enamine

methyl({1-[3-(trifluoromethyl)phenyl]ethyl})amine

  • MDL number MFCD09043460
  • Cas Number : 118761-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33224

Safety Information

msds https://www.enaminestore.com/catalog/EN300-33224
Availability details for
Quantity
  • Availability
  • Add to Cart