ENA321761990 Enamine

5-chloro-1H-indazole-3-carboxylic acid

  • MDL number MFCD02326712
  • Cas Number : 1077-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-34954

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-34954
Availability details for
Quantity
  • Availability
  • Add to Cart