ENA321761992 Enamine

(1R)-1-[3-(trifluoromethyl)phenyl]ethan-1-ol

  • MDL number MFCD02684188
  • Cas Number : 127852-24-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56942
Availability details for
Quantity
  • Availability
  • Add to Cart