ENA321761999 Enamine

2-(1H-1,3-benzodiazol-1-yl)ethan-1-amine

  • MDL number MFCD01050197
  • Cas Number : 55661-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-57356
Availability details for
Quantity
  • Availability
  • Add to Cart