ENA321762003 Enamine

2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}propan-1-amine

  • MDL number MFCD09736423
  • Cas Number : 954270-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57802
Availability details for
Quantity
  • Availability
  • Add to Cart