ENA321762005 Enamine

2-amino-N-[(pyridin-2-yl)methyl]acetamide

  • MDL number MFCD09724217
  • Cas Number : 3014-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57949
Availability details for
Quantity
  • Availability
  • Add to Cart