ENA321762017 Enamine

2-(2-methylpropoxy)ethan-1-amine

  • MDL number MFCD06247813
  • Cas Number : 89585-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-58315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58315
Availability details for
Quantity
  • Availability
  • Add to Cart