ENA321762023 Enamine

(2-methylfuran-3-yl)methanamine

  • MDL number MFCD08435914
  • Cas Number : 35801-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-58810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-58810
Availability details for
Quantity
  • Availability
  • Add to Cart