ENA321762028 Enamine

4-hydrazinyl-6-methylpyrimidine

  • MDL number MFCD12403875
  • Cas Number : 33592-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59247
Availability details for
Quantity
  • Availability
  • Add to Cart