ENA321762033 Enamine

1-(3-aminophenyl)-5-methyl-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD12913281
  • Cas Number : 1211122-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59795
Availability details for
Quantity
  • Availability
  • Add to Cart