ENA321762042 Enamine

5-(2-chloroethyl)-4-methyl-1,3-thiazole

  • MDL number MFCD00868012
  • Cas Number : 533-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60459
Availability details for
Quantity
  • Availability
  • Add to Cart