ENA321762050 Enamine

2,2,2-trifluoroethyl N-[3,5-dimethyl-1-(propan-2-yl)-1H-pyrazol-4-yl]carbamate

  • MDL number MFCD13611096
  • Cas Number : 1209962-44-6
  • Molecular Weight : 279.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-61634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61634
Availability details for
Quantity
  • Availability
  • Add to Cart