ENA321762052 Enamine

1-[4-(1-aminoethyl)phenyl]-5-methyl-2,3-dihydro-1H-imidazol-2-one

  • MDL number MFCD12913285
  • Cas Number : 1209427-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61912
Availability details for
Quantity
  • Availability
  • Add to Cart