ENA321762055 Enamine

4-(1H-imidazol-1-yl)butanoic acid hydrochloride

  • MDL number MFCD11506466
  • Cas Number : 74706-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62061
Availability details for
Quantity
  • Availability
  • Add to Cart