ENA321762059 Enamine

3-(4-ethyl-1,3-thiazol-2-yl)piperidine

  • MDL number MFCD09891526
  • Cas Number : 1017250-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62607
Availability details for
Quantity
  • Availability
  • Add to Cart