ENA321762066 Enamine

2-bromo-1-(3-methylphenyl)ethan-1-one

  • MDL number MFCD07364269
  • Cas Number : 51012-64-7
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47-48
Purity 95
Storage temp. -10
Physical Form oil
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39719

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-39719
pictograms GHS05,GHS07
hstatements H302,H312,H314,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart