ENA321762074 Enamine

2-amino-2-ethylbutan-1-ol

  • MDL number MFCD12827168
  • Cas Number : 19792-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64146

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64146
Availability details for
Quantity
  • Availability
  • Add to Cart