ENA321762082 Enamine

tert-butyl N-[1-(N'-hydroxycarbamimidoyl)propan-2-yl]carbamate

  • MDL number MFCD16833011
  • Cas Number : 1251422-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96...98
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64683

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64683
Availability details for
Quantity
  • Availability
  • Add to Cart