ENA321762083 Enamine

4-amino-N,N-dimethylpyridine-2-carboxamide

  • MDL number MFCD12189433
  • Cas Number : 1155076-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-64783

Safety Information

msds https://www.enaminestore.com/catalog/EN300-64783
Availability details for
Quantity
  • Availability
  • Add to Cart