ENA321762086 Enamine

7-chloro-2,3,4,5-tetrahydro-1-benzoxepin-5-one

  • MDL number MFCD01648758
  • Cas Number : 55579-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-65886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-65886
Availability details for
Quantity
  • Availability
  • Add to Cart