ENA321762105 Enamine

4-(chloromethyl)-5-methyl-1H-imidazole hydrochloride

  • MDL number MFCD11983252
  • Cas Number : 51605-33-5
  • Molecular Weight : 167.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67577
Availability details for
Quantity
  • Availability
  • Add to Cart