ENA321762111 Enamine

1-(2-nitrophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08696938
  • Cas Number : 1006442-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-42130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42130
Availability details for
Quantity
  • Availability
  • Add to Cart