ENA321762121 Enamine

1-[5-fluoro-2-(morpholin-4-yl)phenyl]ethan-1-amine

  • MDL number MFCD11137041
  • Cas Number : 1019627-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69806
Availability details for
Quantity
  • Availability
  • Add to Cart