ENA321762122 Enamine

4,5-dimethyl-2,3-dihydro-1,3-thiazol-2-one

  • MDL number MFCD11632175
  • Cas Number : 25444-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70035

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70035
Availability details for
Quantity
  • Availability
  • Add to Cart