ENA321762124 Enamine

2-(propan-2-yl)pyrimidin-4-ol

  • MDL number MFCD11046847
  • Cas Number : 68210-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70079

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70079
Availability details for
Quantity
  • Availability
  • Add to Cart