ENA321762129 Enamine

oxolan-3-one

  • MDL number MFCD07778393
  • Cas Number : 22929-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Flash Point 56
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70361

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-70361
pictograms GHS02,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart