ENA321762136 Enamine

5-bromo-1H-indazole-3-carboxylic acid

  • MDL number MFCD05663979
  • Cas Number : 1077-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70553

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70553
Availability details for
Quantity
  • Availability
  • Add to Cart