ENA321762139 Enamine

3-[(4-ethoxyphenyl)carbonyl]-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD09932101
  • Cas Number : 1105643-62-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-70784

Safety Information

msds https://www.enaminestore.com/catalog/EN300-70784
Availability details for
Quantity
  • Availability
  • Add to Cart