ENA321762145 Enamine

5-methyl-2-(piperazin-1-yl)-1,3-benzoxazole

  • MDL number MFCD11053076
  • Cas Number : 199292-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70921
Availability details for
Quantity
  • Availability
  • Add to Cart