ENA321762159 Enamine

1-[1-(3-ethyl-1,2,4-oxadiazol-5-yl)ethyl]piperazine

  • MDL number MFCD16705426
  • Cas Number : 1251101-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71047
Availability details for
Quantity
  • Availability
  • Add to Cart