ENA321762164 Enamine

1-[(5-chloro-1-methyl-1H-imidazol-2-yl)methyl]-1,4-diazepane

  • MDL number MFCD11182309
  • Cas Number : 1094226-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71059

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71059
Availability details for
Quantity
  • Availability
  • Add to Cart