ENA321762167 Enamine

4-methoxy-6-methyl-2-(piperazin-1-yl)pyrimidine

  • MDL number MFCD14707799
  • Cas Number : 163613-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71090
Availability details for
Quantity
  • Availability
  • Add to Cart