ENA321762168 Enamine

4-ethoxy-2-(piperazin-1-yl)pyrimidine

  • MDL number MFCD14707783
  • Cas Number : 59215-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71091
Availability details for
Quantity
  • Availability
  • Add to Cart