ENA321762170 Enamine

3,3-dimethyl-1-(piperazin-1-yl)butan-2-ol

  • MDL number MFCD05228423
  • Cas Number : 634576-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71095
Availability details for
Quantity
  • Availability
  • Add to Cart