ENA321762171 Enamine

1-(furan-2-yl)-2-(piperazin-1-yl)ethan-1-ol

  • MDL number MFCD05228228
  • Cas Number : 634559-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71096
Availability details for
Quantity
  • Availability
  • Add to Cart