ENA321762174 Enamine

1-butyl-2-(4-methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06382826
  • Cas Number : 851721-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-18491
Availability details for
Quantity
  • Availability
  • Add to Cart