ENA321762176 Enamine

tert-butyl N-(2-amino-1-cyclopropylethyl)carbamate

  • MDL number MFCD16664908
  • Cas Number : 1250196-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-44010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-44010
Availability details for
Quantity
  • Availability
  • Add to Cart