ENA321762178 Enamine

4-methyl-N1-propylbenzene-1,2-diamine

  • MDL number MFCD11645460
  • Cas Number : 475489-74-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45052
Availability details for
Quantity
  • Availability
  • Add to Cart