ENA321762181 Enamine

3-ethyl-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD08272089
  • Cas Number : 791777-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71127

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71127
Availability details for
Quantity
  • Availability
  • Add to Cart