ENA321762182 Enamine

3-(propan-2-yl)-5H,6H,7H,8H-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD09743507
  • Cas Number : 952182-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71128
Availability details for
Quantity
  • Availability
  • Add to Cart