ENA321762186 Enamine

1-(pyrrolidin-2-ylmethyl)-1H-1,2,4-triazole

  • MDL number MFCD10001573
  • Cas Number : 1018170-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71153

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71153
Availability details for
Quantity
  • Availability
  • Add to Cart