ENA321762187 Enamine

2-(1H-pyrazol-1-ylmethyl)piperidine

  • MDL number MFCD16075576
  • Cas Number : 1249393-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71154
Availability details for
Quantity
  • Availability
  • Add to Cart