ENA321762192 Enamine

6-(2-methoxyethoxy)pyridine-3-carboxylic acid

  • MDL number MFCD09812476
  • Cas Number : 894802-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-45391
Availability details for
Quantity
  • Availability
  • Add to Cart