ENA321762197 Enamine

4-chloro-6-(3-methylpiperidin-1-yl)pyrimidine

  • MDL number MFCD13561820
  • Cas Number : 1220036-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71249
Availability details for
Quantity
  • Availability
  • Add to Cart