ENA321762201 Enamine

3-nitro-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD16042602
  • Cas Number : 1247227-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45530

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-45530
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart