ENA321762207 Enamine

3-(4-methyl-1H-pyrazol-1-yl)butan-2-amine

  • MDL number MFCD14626570
  • Cas Number : 1248214-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-71803

Safety Information

msds https://www.enaminestore.com/catalog/EN300-71803
Availability details for
Quantity
  • Availability
  • Add to Cart