ENA321762210 Enamine

4-(pyridin-2-yloxy)aniline

  • MDL number MFCD06825428
  • Cas Number : 76471-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-71835

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71835
Availability details for
Quantity
  • Availability
  • Add to Cart