ENA321762212 Enamine

2-(1H-1,3-benzodiazol-2-yl)-4-chloroaniline

  • MDL number MFCD00829970
  • Cas Number : 10173-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-71909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-71909
Availability details for
Quantity
  • Availability
  • Add to Cart