ENA321762213 Enamine

3-{[(4-fluorophenyl)methyl]amino}benzamide

  • MDL number MFCD11159266
  • Cas Number : 1042534-72-4
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-72017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-72017
Availability details for
Quantity
  • Availability
  • Add to Cart